ENA413172086 Enamine

1-[(pyrrolidin-2-yl)methyl]azepane

  • MDL number MFCD06653331
  • Cas Number : 881040-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-157152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157152
Availability details for
Quantity
  • Availability
  • Add to Cart