ENA413172087 Enamine

1-iodo-2-methoxy-2-methylpropane

  • MDL number MFCD24459133
  • Cas Number : 19752-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157153
Availability details for
Quantity
  • Availability
  • Add to Cart