ENA413172095 Enamine

4-bromo-3-chloro-N-methyl-N-(propan-2-yl)benzamide

  • MDL number MFCD23030039
  • Cas Number : 1542894-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157227

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157227
Availability details for
Quantity
  • Availability
  • Add to Cart