ENA413172103 Enamine

2-(2-fluorophenoxy)-N'-hydroxyethanimidamide

  • Cas Number : 1016817-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157274
Availability details for
Quantity
  • Availability
  • Add to Cart