ENA413172112 Enamine

1-methyl-5-(trifluoromethyl)-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD27920344
  • Cas Number : 1443278-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157298
Availability details for
Quantity
  • Availability
  • Add to Cart