ENA413172116 Enamine

4-[(4-bromophenyl)methoxy]phenol

  • MDL number MFCD11181705
  • Cas Number : 54589-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157304
Availability details for
Quantity
  • Availability
  • Add to Cart