ENA413172119 Enamine

tert-butyl 4,4-difluoro-2-(hydroxymethyl)pyrrolidine-1-carboxylate

  • MDL number MFCD16250686
  • Cas Number : 317354-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157318
Availability details for
Quantity
  • Availability
  • Add to Cart