ENA413172120 Enamine

2-(2-fluoro-4-methoxyphenyl)acetic acid

  • MDL number MFCD06800744
  • Cas Number : 883531-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157319

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-157319
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart