ENA413172122 Enamine

2-methyl-4-(pyrrolidin-2-yl)-1,3-thiazole

  • MDL number MFCD18646433
  • Cas Number : 1083368-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155348
Availability details for
Quantity
  • Availability
  • Add to Cart