ENA413172124 Enamine

4-chloropyrimidine-2-carbonitrile

  • MDL number MFCD09607718
  • Cas Number : 898044-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157326

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157326
Availability details for
Quantity
  • Availability
  • Add to Cart