ENA413172126 Enamine

2-chloro-5-fluoro-4-methoxy-6-methylpyrimidine

  • MDL number MFCD16621230
  • Cas Number : 1192479-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157340

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157340
Availability details for
Quantity
  • Availability
  • Add to Cart