ENA413172127 Enamine

{5H,6H,7H-cyclopenta[c]pyridazin-3-yl}hydrazine

  • MDL number MFCD16871637
  • Cas Number : 122001-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155349
Availability details for
Quantity
  • Availability
  • Add to Cart