ENA413172128 Enamine

1-[3-fluoro-5-(trifluoromethyl)phenyl]cyclopropane-1-carboxylic acid

  • MDL number MFCD11036981
  • Cas Number : 1260780-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157359

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157359
Availability details for
Quantity
  • Availability
  • Add to Cart