ENA413172135 Enamine

4-methoxy-4-methylpent-1-yn-3-amine

  • MDL number MFCD21822751
  • Cas Number : 1517763-37-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157379
Availability details for
Quantity
  • Availability
  • Add to Cart