ENA413172138 Enamine

2-chloro-4-(propan-2-yloxy)aniline

  • MDL number MFCD16680491
  • Cas Number : 1071989-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157391

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157391
Availability details for
Quantity
  • Availability
  • Add to Cart