ENA413172146 Enamine

tert-butyl 2-(methylcarbamoyl)pyrrolidine-1-carboxylate

  • MDL number MFCD20760962
  • Cas Number : 88815-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157426

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157426
Availability details for
Quantity
  • Availability
  • Add to Cart