ENA413172149 Enamine

(5-amino-1,3,4-oxadiazol-2-yl)methanol

  • MDL number MFCD19206765
  • Cas Number : 23982-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155372
Availability details for
Quantity
  • Availability
  • Add to Cart