ENA413172163 Enamine

4-methoxyadamantane-1-carboxylic acid

  • Cas Number : 2034153-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154906
Availability details for
Quantity
  • Availability
  • Add to Cart