ENA413172170 Enamine

1-{3-methyl-[1,2,4]triazolo[4,3-a]pyrazin-8-yl}piperazine

  • MDL number MFCD01553873
  • Cas Number : 305865-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157509

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157509
Availability details for
Quantity
  • Availability
  • Add to Cart