ENA413172172 Enamine

5-(1-phenyl-1H-pyrazol-4-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD23625227
  • Cas Number : 1500627-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157511

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157511
Availability details for
Quantity
  • Availability
  • Add to Cart