ENA413172173 Enamine

1-[2-chloro-5-(trifluoromethyl)phenyl]cyclopropane-1-carboxylic acid

  • MDL number MFCD11036989
  • Cas Number : 1260676-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157512

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157512
Availability details for
Quantity
  • Availability
  • Add to Cart