ENA413172174 Enamine

2-amino-6H,7H,8H,9H,11H-pyrido[2,1-b]quinazolin-11-one

  • MDL number MFCD24368268
  • Cas Number : 70934-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157513

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157513
Availability details for
Quantity
  • Availability
  • Add to Cart