ENA413172175 Enamine

methyl 4-amino-5-bromopyrimidine-2-carboxylate

  • MDL number MFCD21745467
  • Cas Number : 1504062-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157514
Availability details for
Quantity
  • Availability
  • Add to Cart