ENA413172177 Enamine

2-[(3-phenylpropyl)sulfanyl]ethan-1-ol

  • MDL number MFCD12185602
  • Cas Number : 5755-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157528

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157528
Availability details for
Quantity
  • Availability
  • Add to Cart