ENA413172179 Enamine

2-bromo-6-(2-methoxyethoxy)pyridine

  • MDL number MFCD19619751
  • Cas Number : 1289097-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157531
Availability details for
Quantity
  • Availability
  • Add to Cart