ENA413172180 Enamine

2-(bromomethyl)-3,5-dichloropyrazine

  • MDL number MFCD22376083
  • Cas Number : 1803606-92-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155408

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155408
Availability details for
Quantity
  • Availability
  • Add to Cart