ENA413172181 Enamine

1-(2-fluoro-3-methoxyphenyl)piperazine

  • MDL number MFCD11872745
  • Cas Number : 1121613-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157546
Availability details for
Quantity
  • Availability
  • Add to Cart