ENA413172188 Enamine

3-chloro-1-(1,2,3,4-tetrahydroisoquinolin-2-yl)propan-1-one

  • MDL number MFCD02973833
  • Cas Number : 10579-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157553

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-157553
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart