ENA413172191 Enamine

ethyl 3-(aminomethyl)benzoate hydrochloride

  • MDL number MFCD16660252
  • Cas Number : 91843-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155409

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155409
Availability details for
Quantity
  • Availability
  • Add to Cart