ENA413172195 Enamine

potassium trifluoro(methoxymethyl)boranuide

  • MDL number MFCD10566517
  • Cas Number : 910251-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-155001

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-155001
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart