ENA413172197 Enamine

N-(6-fluoro-1,3-benzothiazol-2-yl)-2-[(4-phenyl-1H-imidazol-2-yl)sulfanyl]acetamide

  • MDL number MFCD04475016
  • Cas Number : 731781-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155011

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155011
Availability details for
Quantity
  • Availability
  • Add to Cart