ENA413172198 Enamine

2,3,4-trimethoxybenzene-1-sulfonamide

  • MDL number MFCD18897552
  • Cas Number : 860514-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157623
Availability details for
Quantity
  • Availability
  • Add to Cart