ENA413172203 Enamine

[4-(trifluoromethyl)-1,3-thiazol-2-yl]methanol

  • MDL number MFCD09909578
  • Cas Number : 204319-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157634

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-157634
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart