ENA413172215 Enamine

5-acetamido-2-methylbenzoic acid

  • MDL number MFCD09260822
  • Cas Number : 103204-70-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-157702

Safety Information

msds https://www.enaminestore.com/catalog/EN300-157702
Availability details for
Quantity
  • Availability
  • Add to Cart