ENA413172219 Enamine

3-[3-(trifluoromethyl)phenoxy]aniline

  • MDL number MFCD06656963
  • Cas Number : 625106-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155421

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155421
Availability details for
Quantity
  • Availability
  • Add to Cart