ENA413172225 Enamine

8-chloro-3-(propan-2-yl)-[1,2,4]triazolo[4,3-a]pyrazine

  • MDL number MFCD20303915
  • Cas Number : 140910-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155018

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155018
Availability details for
Quantity
  • Availability
  • Add to Cart