ENA413172231 Enamine

6-methylmorpholin-3-one

  • MDL number MFCD17292771
  • Cas Number : 127958-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155448

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155448
Availability details for
Quantity
  • Availability
  • Add to Cart