ENA413172259 Enamine

2-amino-3-carbamoylpropanoic acid

  • MDL number MFCD00064400
  • Cas Number : 3130-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-235
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-20760

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-20760
Availability details for
Quantity
  • Availability
  • Add to Cart