ENA413172287 Enamine

N-(2-tert-butylphenyl)-2-chloropropanamide

  • MDL number MFCD12147724
  • Cas Number : 1156595-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27925

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27925
Availability details for
Quantity
  • Availability
  • Add to Cart