ENA413172302 Enamine

1-[6-(1H-imidazol-1-yl)pyridin-3-yl]methanamine

  • MDL number MFCD08235200
  • Cas Number : 914637-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30357
Availability details for
Quantity
  • Availability
  • Add to Cart