ENA413172304 Enamine

N-(2-methoxyethyl)-1,3-benzothiazol-2-amine

  • MDL number MFCD08444687
  • Cas Number : 923688-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30381
Availability details for
Quantity
  • Availability
  • Add to Cart