ENA413172313 Enamine

1'-methyl-3,4-dihydrospiro[1-benzopyran-2,4'-piperidine]-4-amine

  • MDL number MFCD06362451
  • Cas Number : 70505-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30594
Availability details for
Quantity
  • Availability
  • Add to Cart