ENA413172320 Enamine

(E)-N-[(5-chlorofuran-2-yl)methylidene]hydroxylamine

  • MDL number MFCD10606964
  • Cas Number : 54376-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30991

Safety Information

msds https://www.enaminestore.com/catalog/EN300-30991
Availability details for
Quantity
  • Availability
  • Add to Cart