ENA413172322 Enamine

3-[(oxolan-2-yl)methoxy]aniline

  • MDL number MFCD06801284
  • Cas Number : 883536-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31029
Availability details for
Quantity
  • Availability
  • Add to Cart