ENA413172386 Enamine

2-(7-bromo-2,2-dimethyl-2,3-dihydro-1-benzofuran-5-yl)acetonitrile

  • MDL number MFCD27920385
  • Cas Number : 1781146-88-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-43530

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43530
Availability details for
Quantity
  • Availability
  • Add to Cart