ENA413172388 Enamine

2-(7-chloro-2-methyl-2,3-dihydro-1-benzofuran-5-yl)acetonitrile

  • MDL number MFCD27920387
  • Cas Number : 57899-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-43532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43532
Availability details for
Quantity
  • Availability
  • Add to Cart