ENA413172391 Enamine

N-{1-[(2-bromophenyl)methyl]-1H-pyrazol-5-yl}-2-chloroacetamide

  • MDL number MFCD26936307
  • Cas Number : 1803597-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46589

Safety Information

msds https://www.enaminestore.com/catalog/EN300-46589
Availability details for
Quantity
  • Availability
  • Add to Cart