ENA413172399 Enamine

2-(7-bromo-1,3-dioxaindan-5-yl)acetonitrile

  • MDL number MFCD27920397
  • Cas Number : 855937-55-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54744
Availability details for
Quantity
  • Availability
  • Add to Cart