ENA413172417 Enamine

3-methylcyclohexan-1-amine

  • MDL number MFCD00001494
  • Cas Number : 6850-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-33155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33155
Availability details for
Quantity
  • Availability
  • Add to Cart