ENA413172428 Enamine

2-(methylamino)-N-[4-(morpholin-4-yl)phenyl]acetamide

  • MDL number MFCD04632087
  • Cas Number : 875156-12-0
  • Molecular Weight : 249.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(methylamino)-N-[4-(4-morpholinyl)phenyl]acetamide
Inchi Code 1S/C13H19N3O2/c1-14-10-13(17)15-11-2-4-12(5-3-11)16-6-8-18-9-7-16/h2-5,14H,6-10H2,1H3,(H,15,17)
Inchi Key OXAKCURZNNZIAR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-32003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-32003
Availability details for
Quantity
  • Availability
  • Add to Cart